missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human TRAIL (TNFSF10) (soluble) Recombinant Protein, PeproTech®
E. coli Recombinant Protein
80.00€ - 1484.00€
Spécification
Numéro d’adhésion | P50591 |
---|---|
À utiliser avec (application) | ELISA, Functional Assay, Western Blot |
Formule | protein with no preservative |
Identification génétique (Entrez) | 8743 |
Poids moléculaire | 19.6 |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
17862603
|
Gibco™
310-04-100UG |
100 μg |
297.00€
100µg |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
17872203
|
Gibco™
310-04-10UG |
10 μg |
80.00€
10µg |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
17852323
|
Gibco™
310-04-1MG |
1 mg |
1484.00€
1mg |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
17812103
|
Gibco™
310-04-250UG |
250 μg |
520.00€
250µg |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
17882603
|
Gibco™
310-04-500UG |
500 μg |
890.00€
500µg |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
17892023
|
Gibco™
310-04-50UG |
50 μg |
194.00€
50µg |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
Description
TRAIL is a cytokine that belongs to the tumor necrosis factor (TNF) ligand family. TRAIL preferentially induces apoptosis in transformed and tumor cells, but does not appear to kill normal cells although it is expressed at a significant level in most normal tissues. TRAIL binds to several members of TNF receptor superfamily including TNFRSF10A/TRAILR1, TNFRSF10B/TRAILR2, TNFRSF10C/TRAILR3, TNFRSF10D/TRAILR4, and possibly also to TNFRSF11B/OPG. The activity of this protein may be modulated by binding to the decoy receptors TNFRSF10C/TRAILR3, TNFRSF10D/TRAILR4, and TNFRSF11B/OPG that cannot induce apoptosis. The binding of TRAIL protein to its receptors has been shown to trigger the activation of MAPK8/JNK, caspase 8, and caspase 3. Receptors for TRAIL include two death domain containing receptors, death receptor 4 (DR4) and DR5, as well as two decoy receptors, DcR1 and DcR2, lacking the intracellular signaling death domain. TRAIL is a type II membrane protein and expressed in a variety of human tissues. Like TNF and Fas ligand, TRAIL induces apoptosis and NF-kB activation in many tissues and cells.Spécification
P50591 | |
protein with no preservative | |
19.6 | |
E. coli | |
RUO | |
A330042I21Rik; AI448571; Apo-2 ligand; APO2L; Apo-2 L; CD253; CD253 antigen; chemokine tumor necrosis factor ligand superfamily member 10; Ly81; Protein TRAIL; TL2; TN; TNF superfamily member 10; TNF-related apoptosis inducing ligand; TNF-related apoptosis inducing ligand TRAIL; TNF-related apoptosis-inducing ligand; Tnfsf10; TNLG6A; TRAIL; TRAIL protein; TRAIL/APO2L; tumor necrosis factor (ligand) family, member 10; tumor necrosis factor (ligand) superfamily member 10; tumor necrosis factor (ligand) superfamily, member 10; tumor necrosis factor apoptosis-inducing ligand splice variant delta; tumor necrosis factor ligand 6 A; tumor necrosis factor ligand superfamily member 10; tumor necrosis factor superfamily member 10 | |
Human | |
Recombinant | |
Lyophilized |
ELISA, Functional Assay, Western Blot | |
8743 | |
Human TRAIL (TNFSF10) (soluble) | |
-20°C | |
<1 EU/μg | |
TNFSF10 | |
Unconjugated | |
MRERGPQRVA AHITGTRGRS NTLSSPNSKN EKALGRKINS WESSRSGHSF LSNLHLRNGE LVIHEKGFYY IYSQTYFRFQ EEIKENTKND KQMVQYIYKY TSYPDPILLM KSARNSCWSK DAEYGLYSIY QGGIFELKEN DRIFVSVTNE HLIDMDHEAS FFGAFLVG | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit