missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human TNXB (aa 55-173) Control Fragment Recombinant Protein
Click to view available options
Quantité:
100 μl
Conditionnement:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (92%), Rat (92%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-144719 (PA5-144719. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Tenascin-X (TNXB) is a extracellular matrix glycoprotein predominantly located in the outer reticular lamina of the basement membrane (BM). It interacts with many other ECM proteins and it accelerates collagen fibrillogenesisin vitro. TNXB plays a role in interactions between cell and ECM, antiadhesive effect, inhibiting cell migration, and in maintaining homeostasis of the extracellular matrix. Deficiency of TNXB has been associated with the connective tissue disorder Ehlers-Danlos syndrome.
Spécification
Spécification
| Numéro d’adhésion | P22105 |
| Concentration | ≥5.0 mg/mL |
| À utiliser avec (application) | Blocking Assay, Control |
| Formule | 1 M urea, PBS with no preservative; pH 7.4 |
| Identification génétique (Entrez) | 7148 |
| Nom | Human TNXB (aa 55-173) Control Fragment |
| Quantité | 100 μl |
| État réglementaire | RUO |
| Alias de gène | EDS3; growth-inhibiting protein 45; hexabrachion-like protein; HXBL; tenascin XB; tenascin XB1; tenascin XB2; tenascin-X; TENX; Tn-mhc; Tnx; TN-X; TNXB; TNXB1; TNXB2; TNXB2tenascin XB1; TNXBS; TNXBSTNXTENX; VUR8; XB; XBS; XB-S; XBTN-X |
| Nom usuel | TNXB |
| Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu