missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Thrombin ELISA Kit
For the quantitative detection of Human Thrombin
Marque: Invitrogen™ EEL053
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Coagulation factor II is proteolytically cleaved to form thrombin in the first step of the coagulation cascade which ultimately results in the stemming of blood loss. F2 also plays a role in maintaining vascular integrity during development and postnatal life. Mutations in F2 leads to various forms of thrombosis and dysprothrombinemia.Spécification
P00734 | |
0.38 ng/mL | |
ELISA | |
Lecteur de microplaques colorimétriques | |
F2 | |
<10% | |
HRP | |
RUO | |
Refer to product documentation for component specific storage temperature. | |
1 h 20 min |
0.63 to 40 ng/mL | |
HRP | |
Plasma | |
2147 | |
<10% | |
Pre-coated 96 well plate, Standard, Biotinylated Detection Antibody, HRP Conjugate, Standard & Sample Diluent, Biotinylated Detection Antibody Diluent, HRP Conjugate Diluent, Wash Buffer, TMB Substrate, Stop Solution, Plate Sealer | |
96 dosages | |
100 µL | |
Human | |
3 h 30 min |