missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TGR5/GPBAR1 APC-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Marque: R&D Systems FAB4286A
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
TGR5/GPBAR1 Monoclonal specifically detects TGR5/GPBAR1 in Human samples. It is validated for Flow Cytometry.Spécification
TGR5/GPBAR1 | |
Monoclonal | |
APC | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
BG37, BG37hBG37, G protein-coupled bile acid receptor 1, GPBA, GPBAR1, GPCR19, GPR131, G-protein coupled bile acid receptor 1, G-protein coupled bile acid receptor BG37, G-protein coupled receptor GPCR19, M-BAR, M-BARhGPCR19, membrane bile acid receptor, Membrane-type receptor for bile acids, MGC40597, TGR5GPCR | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
151306 | |
Human | |
Purified |
Flow Cytometry | |
409522 | |
Flow Cytometry 10 uL/10^6 cells | |
Q8TDU6 | |
GPBAR1 | |
NS0 mouse myeloma cell line transfected with human TGR5/GPBAR1 Met1-Asn330 Accession # Q8TDU6 | |
100 Tests | |
Primary | |
Stains human TGR5/GPBAR1 transfectants but not irrelevant transfectants in flow cytometry. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2b |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu