missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SERPINB10 Partial ORF (NP_005015.1, 46 a.a. - 145 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marque: Abnova™ H00005273-Q01.10ug
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Sequence: AKGTTAAQMAQVLQFNRDQGVKCDPESEKKRKMEFNLSNSEEIHSDFQTLISEILKPNDDYLLKTANAIYGEKTYAFHNKYLEDMKTYFGAEPQPVNFVESpécification
NP_005015.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
AKGTTAAQMAQVLQFNRDQGVKCDPESEKKRKMEFNLSNSEEIHSDFQTLISEILKPNDDYLLKTANAIYGEKTYAFHNKYLEDMKTYFGAEPQPVNFVE | |
RUO | |
SERPINB10 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
5273 | |
SERPINB10 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PI10/bomapin | |
SERPINB10 | |
Recombinant | |
wheat germ expression system |