Learn More
Invitrogen™ Human RAR gamma-2 (aa 13-28) Synthetic Peptide
Synthetic Peptide
Marque: Invitrogen™ PEP229
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Human RAR gamma 2 (residues 13-28) Synthetic Peptide, PEP-229, from Invitrogen.
PEP-229 is a 16 amino acid synthetic peptide corresponding to amino acid residues 13-28 from human retinoic acid receptor gamma 2. The sequence of this peptide is (amino to carboxy terminus): R (13) R L Y G A A G P G A G L L R R (28).This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product, catalog # PA3-814. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∼70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 ml of distilled water.Spécification
5916 | |
purified | |
Human RAR gamma-2 (aa 13-28) | |
-20°C. Avoid freeze-thaw cycles. | |
Lyophilized | |
Control | |
Yes | |
Peptide | |
RUO - research use only |
Human | |
P13631 | |
0.5 mg/mL | |
Synthetic Peptide | |
peptide with no preservative | |
RARG | |
No preservative | |
50 μg | |
R(13) R L Y G A A G P G A G L L R R(28) |