missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Protocadherin-15 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Marque: R&D Systems MAB6729
300.00 EUR valable jusqu'au 2025-03-28
Utilisez le code promo "25306" pour bénéficier de cette offre.
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Protocadherin-15 Monoclonal specifically detects Protocadherin-15 in Human samples. It is validated for Immunocytochemistry.
Spécification
Protocadherin-15 | |
Monoclonal | |
LYOPH | |
Immunocytochemistry 8-25 ug/mL | |
Q96QU1 | |
PCDH15 | |
Chinese hamster ovary cell line CHO-derived recombinant human Protocadherin-15 Tyr28-Ala1376 Accession # Q96QU1 | |
100 μg | |
Primary | |
Detects human Protocadherin-15 in direct ELISAs. | |
Human | |
Purified |
Immunocytochemistry | |
740523 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
autosomal recessive 23, cadherin-related family member 15, CDHR15, DFNB23, DKFZp667A1711, PCDH15, protocadherin-15, Protocadherin15, protocadherin-related 15, USH1F | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
65217 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Human Protocadherin-15 Antibody, R&D Systems™ > 100ug; Unlabeled
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu