Learn More
Human Pro-BNP Antibody, R&D Systems™
Rat Monoclonal Antibody
Marque: R&D Systems MAB36041
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
BNP Monoclonal specifically detects BNP in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.
Spécification
| BNP | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| P16860 | |
| NPPB | |
| E. coli-derived recombinant human BNP His27-His134 Accession # P16860 | |
| 100 μg | |
| Primary | |
| Detects human Pro-BNP in direct ELISAs. Does not detect BNP-32 (aa 103-134). | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| 370124 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BNF, BNP, brain type natriuretic peptide, Gamma-brain natriuretic peptide, natriuretic peptide B, natriuretic peptide precursor B, natriuretic peptides B, natriuretic protein, NPPB | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4879 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |