missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human NF2/Merlin Antibody, R&D Systems™
Mouse Monoclonal Antibody
Marque: R&D Systems MAB5616
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
NF2/Merlin Monoclonal specifically detects NF2/Merlin in Human samples. It is validated for Western Blot.Spécification
NF2/Merlin | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL | |
P35240 | |
NF2 | |
E. coli-derived recombinant human NF2/Merlin Met363-Lys578 Accession # P35240 | |
100 μg | |
Primary | |
Detects human NF2/Merlin in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human NF1 is observed. | |
Human | |
Purified |
Western Blot | |
534428 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
ACN, BANF, Merlin, moesin-ezrin-radixin like, Moesin-ezrin-radixin-like protein, moesin-ezrin-radizin-like protein, neurofibromin 2 (bilateral acoustic neuroma), neurofibromin 2 (merlin), neurofibromin-2, SCH, Schwannomerlin, Schwannomin | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
4771 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu