Learn More
Novus Biologicals™ Human Neurophysin II/Arg-vasopressin ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Marque:  Novus Biologicals™ NBP2-62140
Code nomenclature Nacres: NA.84
Informations supplémentaires : Poids : 0.00970kg
Code produit. 18228323
  • 2659.76€ / 5 kits
Expédition estimée: 30-09-2024
pour voir le stock disponible 
Description
Description
The Human Neurophysin II/Arg-vasopressin ELISA Kit (Colorimetric) measures human Neurophysin II/Arg-vasopressin in human serum, human plasma, tissue culture media.
Spécification
Spécification
4.10 - 1,000 pg/ml
ELISA Kit (Colorimetric)
ELISA
5 Kit
Arg8-Vasopressin
2.84 pg/ml
Human Plasma, Human Serum, Tissue Culture Media
Human
Storage of components varies. See protocol for specific instructions.