Learn More
Novus Biologicals™ Human Neurophysin II/Arg-vasopressin ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Marque: Novus Biologicals™ NBP2-62140
Code nomenclature Nacres: NA.84
Informations supplémentaires : Poids : 0.00970kg
Description
The Human Neurophysin II/Arg-vasopressin ELISA Kit (Colorimetric) measures human Neurophysin II/Arg-vasopressin in human serum, human plasma, tissue culture media.Spécification
4.10 - 1,000 pg/ml | |
ELISA Kit (Colorimetric) | |
ELISA | |
5 Kit | |
Arg8-Vasopressin |
2.84 pg/ml | |
Human Plasma, Human Serum, Tissue Culture Media | |
Human | |
Storage of components varies. See protocol for specific instructions. |