missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse SIN3A Antibody, R&D Systems™
Goat Polyclonal Antibody
Marque: R&D Systems AF6115
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.00970kg
Description
SIN3A Polyclonal specifically detects SIN3A in Human, Mouse samples. It is validated for Western Blot.Spécification
SIN3A | |
Polyclonal | |
Unconjugated | |
Q96ST3 | |
SIN3A | |
E. coli-derived recombinant human SIN3A Tyr1187-Pro1273 Accession # Q96ST3 | |
100 μg | |
Primary | |
Detects human and mouse SIN3A in Western blots. | |
Human, Mouse | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
DKFZp434K2235, FLJ90319, Histone deacetylase complex subunit Sin3a, KIAA0700, paired amphipathic helix protein Sin3a, SIN3 homolog A, transcription regulator (yeast), SIN3 homolog A, transcriptional regulator (yeast), transcriptional co-repressor Sin3A, Transcriptional corepressor Sin3a, transcriptional regulator, SIN3A | |
Goat | |
Affinity Purified | |
RUO | |
25942 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |