missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse RBP4/Retinol-Binding Protein 4 Antibody, R&D Systems™

Rat Monoclonal Antibody
Marque: R&D Systems MAB34761
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
RBP4/Retinol-Binding Protein 4 Monoclonal specifically detects RBP4/Retinol-Binding Protein 4 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Spécification
| RBP4/Retinol-Binding Protein 4 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.05-2 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| NP_035385 | |
| RBP4 | |
| Mouse myeloma cell line NS0-derived recombinant mouse RBP4/Retinol-Binding Protein 4 Met1-Leu201 Accession # NP_035385 | |
| 100 μg | |
| Primary | |
| Detects mouse RBP4/Retinol-Binding Protein 4 in direct ELISA and Western Blot. In Western blots, detects human RBP4/Retinol-Binding Protein 4. | |
| Human, Mouse | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 423619 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| interstitial, Plasma retinol-binding protein, retinol binding protein 4, plasma, RetinolBinding Protein 4, retinol-binding protein 4, plasma | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5950 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu