missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat TrkB Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 3 publications
Marque: R&D Systems AF397
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
TrkB Polyclonal specifically detects TrkB in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Flow Cytometry, CyTOF-ready.
Spécification
| TrkB | |
| Polyclonal | |
| Unconjugated | |
| BDNF/NT-3 growth factors receptor, EC 2.7.10, EC 2.7.10.1, GP145-TrkB, Neurotrophic tyrosine kinase receptor type 2, neurotrophic tyrosine kinase, receptor, type 2, NTRK2, trk-B, TrkB tyrosine kinase, TRKBBDNF-tropomyosine receptor kinase B, tyrosine kinase receptor B | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 4915 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Flow Cytometry, CyTOF | |
| LYOPH | |
| Western Blot 0.25-0.5 ug/mL, Simple Western 10 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| NTRK2 | |
| Mouse myeloma cell line NS0-derived recombinant human TrkB | |
| 100 μg | |
| Primary | |
| Detects human TrkB in direct ELISA. Detects human, mouse, and rat TrkB Western blots. In direct ELISA and Western blots, less than 1% cross-reactivity with recombinant human (rh) TrkC, rhNT-3, rhNT-4, rhBDNF, and rh beta-NGF is observed. | |
| Human, Mouse, Rat | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu