missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat PLC-gamma 1 Antibody, R&D Systems™
Rat Monoclonal Antibody
Marque: R&D Systems MAB32881
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
PLC-gamma 1 Monoclonal specifically detects PLC-gamma 1 in Human, Mouse, Rat samples. It is validated for Western Blot.Spécification
PLC-gamma 1 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
P19174 | |
PLCG1 | |
E. coli-derived recombinant human PLC-gamma 1 Phe550-His663 Accession # P19174 | |
100 μg | |
Primary | |
Detects endogenous human, mouse, and rat PLC-gamma 1. | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
479819 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EC 3.1.4.11, gamma 1 (formerly subtype 148), monophosphatidylinositol phosphodiesterase, NCKAP3,1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase gamma-1, phosphatidylinositol phospholipase C, phosphoinositidase C, phosphoinositide phospholipase C, Phosphoinositide phospholipase C-gamma-1, phospholipase C, gamma 1, phospholipase C-148, Phospholipase C-gamma-1, Phospholipase C-II, PLC11-phosphatidylinositol-4,5-bisphosphate phosphodiesterase gamma 1, PLC148, PLC-148, PLCG1, PLCgamma 1, PLC-gamma-1, PLC-II1-phosphatidyl-D-myo-inositol-4,5-bisphosphate, PPLCA, triphosphoinositide phosphodiesterase | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5335 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |