missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Proinsulin Biotinylated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Marque: R&D Systems BAM13361
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Proinsulin Monoclonal specifically detects Proinsulin in Human, Mouse samples. It is validated for Intracellular Staining by Flow Cytometry, Immunocytochemistry.Spécification
Proinsulin | |
Monoclonal | |
LYOPH | |
Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL | |
P01308 | |
INS | |
E. coli-derived recombinant human Proinsulin Phe25-Asn110 Accession # P01308 | |
100 μg | |
Primary | |
Detects human and mouse Proinsulin in direct ELISAs. In direct ELISAs, no cross-reactivity with mature Insulin is observed. | |
Human, Mouse | |
Purified |
Flow Cytometry, Immunocytochemistry | |
253627 | |
Biotin | |
Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. with No Preservative | |
IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3630 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu