missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human KPNA3 Partial ORF (NP_002258, 422 a.a. - 521 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marque: Abnova™ H00003839-Q01.10ug
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The transport of molecules between the nucleus and the cytoplasm in eukaryotic cells is mediated by the nuclear pore complex (NPC), which consists of 60-100 proteins. Small molecules (up to 70 kD) can pass through the nuclear pore by nonselective diffusion while larger molecules are transported by an active process. The protein encoded by this gene belongs to the importin alpha family, and is involved in nuclear protein import. [provided by RefSeq]
Sequence: VKDSQVVQVVLDGLKNILIMAGDEASTIAEIIEECGGLEKIEVLQQHENEDIYKLAFEIIDQYFSGDDIDEDPCLIPEATQGGTYNFDPTANLQTKEFNFSpécification
NP_002258 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
VKDSQVVQVVLDGLKNILIMAGDEASTIAEIIEECGGLEKIEVLQQHENEDIYKLAFEIIDQYFSGDDIDEDPCLIPEATQGGTYNFDPTANLQTKEFNF | |
RUO | |
KPNA3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
3839 | |
KPNA3 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
IPOA4/SRP1/SRP1gamma/SRP4/hSRP1 | |
KPNA3 | |
Recombinant | |
wheat germ expression system |