missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-6R alpha Antibody, R&D Systems™
Mouse Monoclonal Antibody
Marque: R&D Systems MAB2271
368.80 EUR valable jusqu'au 2025-03-28
Utilisez le code promo "25306" pour bénéficier de cette offre.
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
IL-6R alpha Monoclonal specifically detects IL-6R alpha in Human samples. It is validated for Immunocytochemistry.
Spécification
IL-6R alpha | |
Monoclonal | |
LYOPH | |
Immunocytochemistry 8-25 ug/mL | |
P08887 | |
IL6R | |
E. coli-derived recombinant human IL-6 R alpha Arg387-Arg468 Accession # P08887 | |
100 μg | |
Primary | |
Detects human IL-6 R alpha in direct ELISAs. | |
Human | |
Purified |
Immunocytochemistry | |
556704 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD126, gp80, IL-6 R alpha, IL6Q, IL6R, IL6R alpha, IL-6R-1, IL6RA, IL-6RA, IL6RQ, interleukin 6 receptor | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3570 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Human IL-6R alpha Antibody, R&D Systems™ > 100ug; Unlabeled
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu