Learn More
ACROBiosystems Human IL-13 R alpha 1 Protein, His Tag
SPR verified
Marque: ACROBiosystems IL1-H5224-100UG
Informations supplémentaires : Poids : 0.00001kg
Description
Interleukin 13 receptor, alpha 1 is also known as IL13RA1, NR4 and CD213A1 (cluster of differentiation 213A1), The IL13 Rα1 cDNA encodes a 427 amino acid (aa) residue precursor protein with a putative 21 aa residue signal peptide, a 324 aa residue extracellular domain, a 23 aa residue transmembrane region and a 59 aa residue cytoplasmic tail. Human and mouseIL13Rα1 share 76% aa sequence identity. IL13RA1 is a subunit of the interleukin 13 receptor. This subunit forms a receptor complex with IL4 receptor alpha, a subunit shared by IL13 and IL4 receptors. This subunit serves as a primary IL13-binding subunit of the IL13 receptor, and may also be a component of IL4 receptors. This protein has been shown to bind tyrosine kinase TYK2, and thus may mediate the signaling processes that lead to the activation of JAK1, STAT3 and STAT6 induced by IL13 and IL4.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 37.7 kDa. The protein migrates as 47-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spécification
NP_001551.1 | |
100 μg | |
Gly 22 - Thr 343 | |
IL13RA1,CD213A1,IL-13 Ra,NR4,RP13-128O4.2,IL13Rα1 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
37.7 kDa | |
Human IL-13 R alpha 1, His Tag (SPR verified) (IL1-H5224) is expressed from human 293 cells (HEK293). It contains AA Gly 22 - Thr 343 (Accession # NP_001551.1). | |
-20°C | |
IL-13 R alpha 1 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |