missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human IL-12 p80 Recombinant Protein, PeproTech®
Insect cells Recombinant Protein
Marque: Gibco™ 200-12P80H-50UG
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
IL-12 is a disulfide-linked heterodimeric protein (p70), composed of two subunits, p35 and p40, which are encoded by two different genes. Accumulating data indicates that p40 secretion precedes that of IL-12 expression. In addition, to its ability to covalently bind to p35 to form IL-12, p40 can bind to p19 to form IL-23, or it can form the homodimer designated IL-12 p80. Elevated levels of IL-12 p80 correlate to macrophage recruitment and increased inflammation in asthma and respiratory viral infection models.Spécification
P29460 | |
protein with no preservative | |
80 | |
50 μg | |
-20°C | |
<1 EU/μg | |
IL12B | |
Unconjugated | |
p40 subunit: IWELKKDVYV VELDWYPDAP GEMVVLTCDT PEEDGITWTL DQSSEVLGSG KTLTIQVKEF GDAGQYTCHK GGEVLSHSLL LLHKKEDGIW STDILKDQKE PKNKTFLRCE AKNYSGRFTC WWLTTISTDL TFSVKSSRGS SDPQGVTCGA ATLSAERVRG DNKEYEYSVE CQEDSACPAA EESLPIEVMV DAVHKLKYEN YTSSFFIRDI IKPDPPKNLQ LKPLKNSRQV EVSWEYPDTW STPHSYFSLT FCVQVQGKSK REKKDRVFTD KTSATVICRK NASISVRAQD RYYSSSWSEW ASVPCS | |
≥ 95% by SDS-PAGE gel and HPLC analyses. |
Functional Assay | |
3593 | |
Human IL-12 p80 | |
Insect cells | |
RUO | |
IL-12 | |
Human | |
Recombinant | |
Lyophilized |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu