missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-1 alpha/IL-1F1 Fluorescein-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 1 publication
Marque: R&D Systems IC200F
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
IL-1 alpha/IL-1F1 Monoclonal specifically detects IL-1 alpha/IL-1F1 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Spécification
IL-1 alpha/IL-1F1 | |
Monoclonal | |
Fluorescein | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
BAF, Hematopoietin-1, IL1, IL1 alpha, IL-1 alpha, IL1A, IL-1A, IL1-ALPHA, IL1F1, IL-1F1, IL1F1hematopoietin-1, interleukin 1, alpha, interleukin-1 alpha, LAF, LEM, preinterleukin 1 alpha, pro-interleukin-1-alpha | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3552 | |
Human | |
Purified |
Flow Cytometry | |
4414 | |
Intracellular Staining by Flow Cytometry 10 uL/10^6 cells | |
P01583 | |
IL1A | |
E. coli-derived recombinant human IL-1 alpha/IL-1F1 Ser113-Ala271 Accession # P01583 | |
100 Tests | |
Primary | |
Detects human IL-1 alpha/IL-1F1 in ELISAs and Western blots. In ELISAs, this antibody does not cross-react with recombinant human (rh) IL-1 beta , -2, -3, -4, -5, -6, -7, -8, -9, -10, -11, -12, -13, rmIL-1 alpha , -1 beta , -3, -4, -5, -6, -7, -9, or -13. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2a |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Human IL-1 alpha/IL-1F1 Fluorescein-conjugated Antibody, R&D Systems™ > 100 Tests; Fluorescein
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu