missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human IFN-alpha 2 (alpha2a) Recombinant Protein, PeproTech®

Code produit. 17896898
Click to view available options
Quantité:
1 mg
100 μg
20 μg
250 μg
500 μg
Conditionnement:
100µg
1mg
20µg
250µg
500µg
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Code produit. 17896898

Marque: Gibco™ 30002AA1MG

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Recombinant Protein

Recombinant Human IFN-alpha (2a) is a 19.3 kDa protein containing 166 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

IFN alpha (Interferon alpha) is a Type I interferon belonging to a family of structurally related, species-specific proteins exclusive to vertebrates. The interaction of IFN-alpha with its receptor component result in the activation of a number of signaling pathways which are mainly involved in innate immune response against viral infection. In humans, the gene for IFN alpha is located on on chromosome 9. The type I interferons consist of 14 different alpha isoforms (subtypes with slightly different specificities), and single beta, omega, epsilon and kappa isoforms. Homologous molecules are found in many species, including rats and mice (and most mammals), and have been identified in birds, reptiles and fish species. IFN alpha is produced primarily by plasmacytoid dendritic cells, is a potent component of the anti-viral innate immune response and modulates adaptive immunity. Diseases associated with the dysfunction of IFN alpha include Hepatitis C and Hepatitis.
TRUSTED_SUSTAINABILITY

Spécification

Numéro d’adhésion P01563
À utiliser avec (application) Functional Assay
Format Lyophilized
Formule protein with no preservative
Identification génétique (Entrez) 3440
Poids moléculaire 19.3 kDa
Nom Human IFN-alpha 2 (alpha2a)
Méthode de purification HPLC, SDS-PAGE
Quantité 1 mg
État réglementaire RUO
Concentration en endotoxines <1 EU/ μg
Alias de gène alpha-2 A interferon; Ifa2; IFNA; IFN-a; IFNA 2; Ifna2; IFNA2A; IFNA2B; IFNA2C; IFN-alpha 1 b; IFN-alpha-1/13; IFN-alpha-2; IFN-alphaA; INFA2; Interferon; interferon alpha 1 b; interferon alpha 2; interferon alpha 2 A; interferon alpha 2 b; interferon alpha A; interferon alpha family, gene 2; interferon alpha-2; Interferon alpha-A; interferon alpha-D; interferon, alpha 2; Interferon-alpha II; leIF A; leIF D; RP11-354P17.2
Nom usuel Interferon alpha-2
Symbole de gène(s) IFNA2
Activité biologique Determined by a cytotoxicity assay using human TF-1 cells.
Type de produit Protein
Conjugué Unconjugated
Recombinant Recombinant
Séquence MCDLPQTHSL GSRRTLMLLA QMRKISLFSC LKDRHDFGFP QEEFGNQFQK AETIPVLHEM IQQIFNLFST KDSSAAWDET LLDKFYTELY QQLNDLEACV IQGVGVTETP LMKEDSILAV RKYFQRITLY LKEKKYSPCA WEVVRAEIMR SFSLSTNLQE SLRSKE
Contenu et stockage -20°C
Activité Determined by a cytotoxicity assay using human TF-1 cells.
Système d’expression E. coli
Forme Lyophilized
Pureté ou qualité ≥ 98% by SDS-PAGE gel and HPLC analyses.
Afficher plus Afficher moins
Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis