missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human CD257 (BAFF, BLyS) Synthetic Peptide
Synthetic Peptide
Marque: Invitrogen™ PEP0066
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
BAFF Synthetic Peptide, PEP-0066, from Invitrogen.
This peptide corresponds to 16 amino acids near the carboxy terminus of human BAFF.PEP-0066 can be used as a blocking peptide with polyclonal antibody PA5-19941.Spécification
10673 | |
purified | |
Human CD257 (BAFF, BLyS) | |
-20°C | |
Liquid | |
Blocking Assay, Control | |
No | |
Peptide | |
RUO - research use only |
Human | |
Q9Y275 | |
200 μg/mL | |
Synthetic Peptide | |
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 | |
TNFSF13B | |
0.02% sodium azide | |
50 μg | |
PBS |