missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD2 Antibody, R&D Systems™
Goat Polyclonal Antibody
Marque: R&D Systems AF1856-SP
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
CD2 Polyclonal specifically detects CD2 in Human samples. It is validated for Western Blot, Simple Western, Flow Cytometry, CyTOF-ready.Spécification
CD2 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD2, CD2 antigen, CD2 antigen (p50), sheep red blood cell receptor, CD2 molecule, Erythrocyte receptor, FLJ46032, LFA-2, LFA-3 receptor, lymphocyte-function antigen-2, Rosette receptor, SRBC, T11, T-cell surface antigen CD2, T-cell surface antigen T11/Leu-5 | |
Goat | |
Affinity Purified | |
RUO | |
914 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF | |
LYOPH | |
Western Blot 0.5 ug/mL, Simple Western 10 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 3-15 ug/mL, CyTOF-ready | |
P06729 | |
CD2 | |
Mouse myeloma cell line NS0-derived recombinant human CD2 Lys25-Asp209 Accession # P06729 | |
25 μg | |
Primary | |
Detects human CD2 in direct ELISAs and Western blots. | |
Human | |
IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu