missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD19 Alexa Fluor 647-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Marque: R&D Systems FAB4867R-100
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
CD19 Monoclonal specifically detects CD19 in Human samples. It is validated for Flow Cytometry.Spécification
CD19 | |
Monoclonal | |
Alexa Fluor 647 | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
CD19 | |
Human CLL cells | |
100 Tests | |
Primary | |
Detects human CD19. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG1 |
Flow Cytometry | |
4G7-2E3 | |
Flow Cytometry 5 uL/10^6 cells | |
B4, B-lymphocyte antigen CD19, B-lymphocyte surface antigen B4, CD19, CD19 antigen, CD19 molecule, CVID3, Differentiation antigen CD19, Leu-12, MGC12802, T-cell surface antigen Leu-12 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
930 | |
Human | |
Supernatant |
For Research Use Only