Learn More
Invitrogen™ Human CASK (aa 578-592) Synthetic Peptide
Synthetic Peptide
Marque: Invitrogen™ PEP110
Code nomenclature Nacres: NA.26
Informations supplémentaires : CAS : 69-74-9 Poids : 0.01000kg
Description
Human CASK (residues 578-592) Synthetic Peptide, PEP-110, from Invitrogen.
PEP-110 is a 15 amino acid synthetic peptide whose sequence corresponds to amino acid residues 578-592 of human CASK. The sequence of this peptide is (amino to carboxy terminus): C (578)-E-R-D-S-P-S-T-S-R-Q-S-P-A-N (592)This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and human CASK, catalog # PA1-544. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∼70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 ml of distilled water.Spécification
8573 | |
purified | |
Human CASK (aa 578-592) | |
-20°C. Avoid freeze-thaw cycles. | |
Lyophilized | |
Control | |
Yes | |
Peptide | |
RUO - research use only |
Human | |
O14936 | |
0.5 mg/mL | |
Synthetic Peptide | |
peptide with no preservative | |
CASK | |
No preservative | |
50 μg | |
C(578)-E-R-D-S-P-S-T-S-R-Q-S-P-A-N(592) |