missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human BTK Antibody, R&D Systems™
Mouse Monoclonal Antibody
Marque: R&D Systems MAB5807
392.00 EUR valable jusqu'au 2025-03-28
Utilisez le code promo "25306" pour bénéficier de cette offre.
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
BTK Monoclonal specifically detects BTK in Human samples. It is validated for Western Blot, Immunocytochemistry.
Spécification
BTK | |
Monoclonal | |
LYOPH | |
Western Blot 0.05 ug/mL, Immunocytochemistry 8-25 ug/mL | |
Q06187 | |
BTK | |
E. coli-derived recombinant human BTK Glu280-Val377 Accession # Q06187 | |
100 μg | |
Primary | |
Detects endogenous human BTK in Western blots. | |
Human | |
Purified |
Western Blot, Immunocytochemistry | |
566928 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Agammaglobulinaemia tyrosine kinase, AGMX1, AGMX1MGC126262, AT, ATKIMD1, B-cell progenitor kinase, BPK, Bruton agammaglobulinemia tyrosine kinase, Bruton tyrosine kinase, dominant-negative kinase-deficient Brutons tyrosine kinase, EC 2.7.10, EC 2.7.10.2, IMD1, PSCTK1, tyrosine-protein kinase BTK, XLA, XLAMGC126261 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
695 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Human BTK Antibody, R&D Systems™ > 100ug; Unlabeled
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu