missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human 20S Proteasome Z (aa 263-277) Synthetic Peptide
Synthetic Peptide
Marque: Invitrogen™ PEP175
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Human 20S Proteasome Z (residues 263-277) Synthetic Peptide, PEP-175, from Invitrogen.
PEP-175 is a 15 amino acid synthetic peptide whose sequence is from human proteasome 20S Z. The sequence of this peptide is (amino to carboxy terminus): C-E (263)-I-E-V-L-E-E-T-V-Q-T-M-D-T-S (NH2) (277)This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and human proteasome 20S Z, catalog # PA1-979. Using a solution with equal weights per unit volume of peptide and corresponding antibody will yield a solution with a large molar excess of peptide that is able to competitively bind the antibody.Reconstitute with 0.1 ml of distilled water.Spécification
5695 | |
purified | |
Human 20S Proteasome Z (aa 263-277) | |
-20°C. Avoid freeze-thaw cycles. | |
Lyophilized | |
Control | |
Yes | |
Peptide | |
RUO - research use only |
Human | |
Q99436 | |
0.5 mg/mL | |
Synthetic Peptide | |
peptide with no preservative | |
PSMB7 | |
No preservative | |
50 μg | |
C-E(263)-I-E-V-L-E-E-T-V-Q-T-M-D-T-S(NH2)(277) |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu