missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human 11S REG gamma (aa 1-14) Synthetic Peptide
Synthetic Peptide
Marque: Invitrogen™ PEP091
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Human 11S REG gamma (residues 1-14) Synthetic Peptide, PEP-091, from Invitrogen.
PEP-091 is a 14 amino acid synthetic peptide whose sequence corresponds to residues 1-17 of human proteasome activator 11S REG gamma with a carboxy-terminal cysteine added for carrier protein conjugation. The sequence of this peptide is (amino to carboxy terminus): M1-A-S-L-L-K-V-D-Q-E-V-K-L-K (14)-CThis peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and human proteasome activator 11S REG gamma, catalog # PA1-961. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∼70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 ml of distilled water.Spécification
10197 | |
purified | |
Human 11S REG gamma (aa 1-14) | |
-20°C. Avoid freeze-thaw cycles. | |
Lyophilized | |
Control | |
Yes | |
Peptide | |
RUO - research use only |
Human | |
P61289 | |
0.5 mg/mL | |
Synthetic Peptide | |
peptide with no preservative | |
PSME3 | |
No preservative | |
50 μg | |
M1 - A - S - L - L - K - V - D - Q - E - V - K - L - K(14) - C |