missing translation for 'onlineSavingsMsg'
Learn More

hnRNP H Antibody - Azide and BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

231.00€ - 468.00€

Spécification

Antigène hnRNP H
Dilution Western Blot 1:1000 - 1:5000, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugué Unconjugated
Consulter d'autres spécifications

Produits 2
Code produit Marque Quantité Prix Quantité et disponibilité  
Code produit Marque Quantité Prix Quantité et disponibilité  
18626580
Afficher les documents
Novus Biologicals
NBP2-93590-0.02ml
0.02 mL
231.00€
0.02mL
Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!
18621371
Afficher les documents
Novus Biologicals
NBP2-93590-0.1ml
0.1 mL
468.00€
0.10mL
Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!
Description

Description

hnRNP H Polyclonal antibody specifically detects hnRNP H in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Spécification

Spécification

hnRNP H
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Cellular Markers, Translation Control
PBS (pH 7.3), 50% glycerol
3187
IgG
Affinity purified
Western Blot 1:1000 - 1:5000, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin
Polyclonal
Purified
RUO
Human, Mouse, Rat
DKFZp686A15170, heterogeneous nuclear ribonucleoprotein H, heterogeneous nuclear ribonucleoprotein H1 (H), hnRNP H, HNRPH, HNRPH1hnRNPH
Recombinant fusion protein containing a sequence corresponding to amino acids 191-449 of human hnRNP H (NP_005511.1). VRTHYDPPRKLMAMQRPGPYDRPGAGRGYNSIGRGAGFERMRRGAYGGGYGGYDDYNGYNDGYGFGSDRFGRDLNYCFSGMSDHRYGDGGSTFQSTTGHCVHMRGLPYRATENDIYNFFSPLNPVRVHIEIGPDGRVTGEADVEFATHEDAVAAMSKDKANMQHRYVELFLNSTAGASGGAYEHRYVELFLNSTAGASGGAYGSQMMGGMGLSNQSSYGGPASQQLSGGYGGGYGGQSSMSGYDQVLQENSSDFQSNIA
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Vidéos
FDS
Documents

Documents

Product Certifications