Learn More
HMGXB4 Rabbit anti-Human, Proteintech
Rabbit PolyClonal Antibody
Marque: Proteintech 24821-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
High mobility group (HMG) proteins are nonhistone chromosomal proteins.Spécification
HMGXB4 | |
Polyclonal | |
Unconjugated | |
HMGXB4 | |
HMG box containing protein 4, HMG box domain containing 4, HMG2L1, HMGBCG, HMGXB4, Protein HMGBCG, THC211630 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10042 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9UGU5 | |
HMGXB4 | |
HMGXB4 Fusion Protein Ag20424 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |