missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA-DRB5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 21702-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
HLA-DRB5 belongs to the HLA class II beta chain paralogues. This class II molecule is a heterodimer consisting of an alpha (DRA) and a beta (DRB) chain, both anchored in the membrane. It plays a central role in the immune system by presenting peptides derived from extracellular proteins. Class II molecules are expressed in antigen presenting cells (APC: B lymphocytes, dendritic cells, macrophages). The beta chain is approximately 26-28 kDa and its gene contains 6 exons. Exon one encodes the leader peptide, exons 2 and 3 encode the two extracellular domains, exon 4 encodes the transmembrane domain and exon 5 encodes the cytoplasmic tail. Within the DR molecule the beta chain contains all the polymorphisms specifying the peptide binding specificities. Typing for these polymorphisms is routinely done for bone marrow and kidney transplantation. DRB1 is expressed at a level five times higher than its paralogues DRB3, DRB4 and DRB5. The presence of DRB5 is linked with allelic variants of DRB1, otherwise it is omitted. There are 4 related pseudogenes: DRB2, DRB6, DRB7, DRB8 and DRB9.Spécification
HLA-DRB5 | |
Polyclonal | |
Unconjugated | |
HLA-DRB5 | |
DR beta 2, DR beta 5, Dw2, HLA DRB, HLA DRB5, HLA-DRB5, MHC class II antigen DRB5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
3127 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q30154 | |
HLA-DRB5 | |
HLA-DRB5 Fusion Protein Ag16307 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |