missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HDAC2 Recombinant Rabbit Monoclonal Antibody (9H4)

Rabbit Recombinant Monoclonal Antibody
Marque: Invitrogen™ MA538405
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
This antibody has been tested in direct-ELISA.
HDAC2 belongs to the histone deacetylase family. Histone deacetylases act via the formation of large multiprotein complexes and are responsible for the deacetylation of lysine residues at the N-terminal regions of core histones (H2A, H2B, H3 and H4). This protein forms transcriptional repressor complexes by associating with many different proteins, including YY1, a mammalian zinc-finger transcription factor. Thus, it plays an important role in transcriptional regulation, cell cycle progression and developmental events. Alternative splicing results in multiple transcript variants.
Spécification
| HDAC2 | |
| Recombinant Monoclonal | |
| 0.17 mg/mL | |
| PBS with 50% glycerol, 150mM NaCl and 0.02% sodium azide; pH 7.4 | |
| Q92769 | |
| HDAC2 | |
| A synthesized peptide derived from human HDAC2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 9H4 | |
| Unconjugated | |
| HDAC2 | |
| D10Wsu179e; HD2; hdac 2; Hdac2; hdac-2; Histone deacetylase 2; histone deacetylase 2-like protein; histone deacetylase-2; I79_010219; mRPD3; RPD3; transcriptional regulator homolog RPD3; YAF1; YY1 transcription factor-binding protein; YY1-associated factor 1; Yy1bp | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 3066 | |
| -20°C or -80°C if preferred | |
| Liquid |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu