Learn More
Thermo Scientific™ Harvestainer™ Microcarrier Separation System
GREENER_CHOICE
Code produit. 17829796 Tous les produits Thermo Scientific Produits
Click to view available options
Capacité (métrique):
3 l
12 l
Conditionnement:
1 pièce
Code produit. 17829796
Marque: Thermo Scientific™ SH31268.02
The Thermo Scientific Harvestainer Microcarrier Separation System is the ideal solution for the separation of microcarrier beads during adherent cell culture production.
The Thermo Scientific Harvestainer Microcarrier Separation System is the ideal solution for the separation of microcarrier beads during adherent cell culture production. The Harvestainer system is a closed, single-use system that helps to increase product yields versus traditional methods, while reducing clean-in-place and steam-in-place requirements and water for injection usage. It eliminates equipment maintenance and reduces process validations, cycle times, and manual final decanting processes.
For smaller-scale or larger-scale applications, the Harvestainer system is ideal for in-line microcarrier separation. The 3 L and 12 L Harvestainer systems consist of a pre-assembled bioprocess container (BPC) and tray in a single-use unit, where the tray acts as the secondary containment device. The 25 L and 50 L Harvestainer systems consist of a 3D BPC with a drum as the secondary containment device. The BPC features a dip tube with dipwell design for better drainage and minimal manipulation.
• Closed system
• Scalable for both smaller-scale and larger-scale applications
• Helps deliver high recovery rates
• Inline microcarrier separation
Applications
• Adherent cell culture production
TRUSTED_SUSTAINABILITY
Spécification
Capacité (métrique) 12 l
Quantité 12 L
Tubing Style TPE