missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSTP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 15902-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
GSTP1 is a member of the glutathione S-transferases (GSTs), which are a family of enzymes that play an important role in detoxification by catalyzing the conjugation of many hydrophobic and electrophilic compounds with reduced glutathione. Based on their biochemical, immunologic, and structural properties, the soluble GSTs are categorized into 4 main classes: alpha, mu, pi, and theta. This GST family member is a polymorphic gene encoding active, functionally different GSTP1 variant proteins that are thought to function in xenobiotic metabolism and play a role in susceptibility to cancer, and other diseases.Spécification
GSTP1 | |
Polyclonal | |
Unconjugated | |
GSTP1 | |
DFN7, FAEES3, Glutathione S transferase P, glutathione S transferase pi 1, GST class pi, GST3, GSTP1, GSTP1 1, PI | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14870, 24426, 2950 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P04906, P09211, P19157 | |
Gstp1 | |
GSTP1 Fusion Protein Ag8731 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |