missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRAMD1B Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 24905-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
GRAMD1B, also named as KIAA1201, contains a transmembrane region and two domains of known function: the GRAM domain and a VASt domain. It is predicted to localize in the nucleus, supported by several nuclear transport signals and nuclear associated motifs. This highly conserved gene is found in a variety of vertebrates and invertebrates, however is not found in bacteria or fungi. There are some isoforms with MW 50 kDa and 81-87 kDa. With phosphor modification, it is about 90-110 kDa in WB detection.Spécification
GRAMD1B | |
Polyclonal | |
Unconjugated | |
GRAMD1B | |
GRAM domain containing 1B, GRAMD1B, KIAA1201 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
235283, 57476 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q3KR37, Q80TI0 | |
GRAMD1B | |
GRAMD1B Fusion Protein Ag20670 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |