Learn More
BUCHI Cartouches FlashPure Silica Flash
Haute résolution et capacité de charge exceptionnelle
Marque: BUCHI 11067704
Includes: 20 Pieces
Description
- High resolution of separation due to high-quality silica
- Increased fraction purity due to a better separation
- Faster evaporation of smaller fraction volumes
- Reproducible performance from batch to batch
- Increased sample throughput due to high sample loading capacity
- Solvent saving: required purification yield is achieved faster
Spécification
Silica - Normal Phase | |
20 Pc. | |
For Flash Chromatography Using ISCO and Biotage Instruments |
FlashPure | |
12 g | |
75 psig |