Learn More
GOLSYN Mouse anti-Human, Unconjugated, Clone: 3B11C5, Proteintech
Mouse Monoclonal Antibody
Marque:  Proteintech 67595-1-IG-20UL
 Afficher plus de versions de ce produit
Code produit. 16800316
  • 145.00€ / 20µL
Découvrez plus d'offres spéciales
Description
Description
The GOLSYN antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human GOLSYN. This antibody recognizes human antigen. The GOLSYN antibody has been validated for the following applications: WB, ELISA analysis.
Spécification
Spécification
GOLSYN
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, ELISA
3B11C5
Unconjugated
20 μL
55638
IgG2b
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.