missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Human IgG whole molecule, HRP, Polyclonal, Secondary Antibody, Abnova™
Marque: Abnova PAB10692.2mg
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Spécification
| Goat anti-Human IgG whole molecule | |
| Polyclonal | |
| Goat anti-human IgG recognizes human IgG whole molecule. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Peroxidase. | |
| Lyophilized from 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% gentamicLyophilized from sulfate) | |
| Human IgG whole molecule | |
| 2 mg | |
| Human | |
| Lyophilized |
| Dot Blot, ELISA, Immunohistochemistry, Western Blot | |
| HRP | |
| The optimal working dilution should be determined by the end user. | |
| Goat | |
| This product was prepared from monospecific antiserum by immunoaffinity chromatography, followed by solid phase adsorption(s) to remove any unwanted reactivities. | |
| Secondary | |
| Store at 4°C on dry atmosphere. After reconstitution with 1 mL of deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu