missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glut1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Marque: Novus Biologicals NB110-39113
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Glut1 Polyclonal specifically detects Glut1 in Human, Mouse, Rat, Rabbit samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, In vitro assay, Flow (Intracellular).
Spécification
| Glut1 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| Choreoathetosis/Spasticity, Episodic (Paroxysmal Choreoathetosis), CSE, DYT17, DYT18, DYT9, EIG12, Glucose transporter type 1, erythrocyte/brain, GLUT-1, GLUT1DS, HepG2 glucose transporter, HTLVR, Human T-Cell Leukemia Virus (I and II) Receptor, MGC141895, MGC141896, PED, solute carrier family 2 (facilitated glucose transporter), member 1, Solute Carrier Family 2 Member 1, solute carrier family 2, facilitated glucose transporter member 1 | |
| Rabbit | |
| 54.1 kDa | |
| 0.1 mL | |
| Cancer, Cardiovascular Biology, Cellular Markers, Diabetes Research, Lipid and Metabolism, Membrane Trafficking and Chaperones, Neuroscience, Plasma Membrane Markers, Signal Transduction | |
| 6513 | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, ChIP Assay, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), In vitro Assay | |
| 1 mg/mL | |
| Western Blot 1:500, Simple Western 1:50, Chromatin Immunoprecipitation reported in scientific literature, Flow Cytometry 1 ug/ml. Use reported by customer review, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry-Paraffin 1:200, Immunohistochemistry-Frozen 1:200. Use reported in scientific literature, In vitro assay reported in scientific literature (Trachsel V et al), Flow (Intracellular) 1 ug/ml, Chromatin Immunoprecipitation (ChIP) | |
| P11166 | |
| SLC2A1 | |
| This Glut1 antibody is made against a synthetic peptide made to an N-terminal region of the human GLUT1 protein (between residues 1-100). [Swiss-Prot# P11166]. The immunogen is cytosolic. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Reacts with human and mouse. 100% sequence identity with primate protein, 93% sequence identity with rat, cow, and rabbit proteins. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu