Learn More
GCLM Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca:  Proteintech 14241-1-AP-20UL
 vedi altre versioni di questo prodotto
Codice prodotto. 16832794
  • 145.00€ / 20µL
Esplora altre offerte speciali
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Descrizione
Descrizione
Gamma-glutamylcysteine synthetase (gamma-GCS) is the rate limiting enzyme for glutathione (L-gamma-glutamyl-L-cysteinylglycine, GSH) synthesis. GSH is ubiquitous in mammalian cells as a vital intra- and extracellular protective antioxidant. gamma-GCS is a heterodimer of a heavy catalytic subunit and a light regulatory subunit that is responsive to inflammation, phenolic ntioxidants, heat shock, oxidants and cytokines.
Specifica
Specifica
GCLM
Polyclonal
Unconjugated
GCLM
Gamma ECS regulatory subunit, GCLM, GCS light chain, GLCLR
Rabbit
Antigen Affinity Chromatography
RUO
14630, 2730, 29739
-20°C
Liquid
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry, Flow Cytometry
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O09172, P48507, P48508
GCLM
GCLM Fusion Protein Ag5497
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Suggerimenti di prodotto
Suggerimenti di prodotto
Videos
SDS
Documenti
Documenti
Product Certifications
Offerte speciali
Offerte speciali