missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GATA1 Polyclonal Antibody

Goat Polyclonal Antibody
Marque: Invitrogen™ PA518408
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
This antibody is predicted to react with bovine, canine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.
Transcriptional activator or repressor which probably serves as a general switch factor for erythroid development. It binds to DNA sites with the consensus sequence 5'-[AT]GATA[AG]-3' within regulatory regions of globin genes and of other genes expressed in erythroid cells.
Spécification
| GATA1 | |
| Polyclonal | |
| Unconjugated | |
| Gata1 | |
| eryf1; Erythroid transcription factor; erythroid transcription factor 1; GATA binding protein 1; GATA binding protein 1 (globin transcription factor 1); GATA1; GATA-1; GATA-binding factor 1; GATA-binding protein 1; GATA-binding protein 1 (globin transcription factor 1); GF1; Gf-1; globin transcription factor 1; HGNC:4170; NFE1; NF-E1; NF-E1 DNA-binding protein; nuclear factor, erythroid 1; transcription factor GATA1; XLANP; XLTDA; XLTT | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 14460, 2623 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P15976, P17679 | |
| Gata1 | |
| Synthetic peptide sequence (DAEAYRHSPVFQ) corresponding to the internal amino acids of GATA1 (aa 65-76). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction