Learn More
GARP Mouse, APC, Clone: GARP5, enQuireBio™
Mouse Monoclonal Antibody
Marque:  EnquireBio QAB84-APC-25Tests
Code produit. 15982558
  • 250.00€ / 25 tests
Découvrez plus d'offres spéciales
Spécification
GARP
Monoclonal
5μL/Test
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
LRRC32
Protein A/G Purified
RUO
2615
IgG1
Flow Cytometry
GARP5
APC
XP_016873016
Mouse
25 Tests
Primary
Store at 2-4°C.