Learn More
Tocris Bioscience™ FTY 720
Click to view available options
Quantité:
10 mg
50 mg
Conditionnement:
10mg
50mg
Description
FTY 720 is an S1P receptor agonist; undergoes phosphorylation by sphingosine kinase 2, which then binds S1P1,3,4 and 5 receptors (EC50 values are 0.3 - 0.6 nM for S1P1, 4 and 5 and ∼3 nM for S1P3). Immunosuppressant; sequesters lymphocytes in lymph nodes, reducing autoimmune response. Suppresses the development of EAE (an in vivo model of MS). Orally bioavailable and blood-brain barrier permeable.
Spécification
Spécification
| CAS | 162359-56-0 |
| Quantité | 50 mg |
| Cible | Sphingosine-1-phosphate Receptor |
| Formule moléculaire | C19H33NO2.HCl |
| Poids de la formule | 343.93g/mol |
| Type de produit | Agonist |
Usage exclusivement réservé à la recherche