missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FSCN3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 13536-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Acts as an actin bundling protein.Spécification
FSCN3 | |
Polyclonal | |
Unconjugated | |
FSCN3 | |
Fascin 3, FSCN3, Testis fascin | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
296947, 29999, 56223 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9NQT6, Q9QXW4 | |
Fscn3 | |
FSCN3 Fusion Protein Ag4459 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |