Learn More
Labconco FreeZone Stoppering Tray Dryer
Tous les produits Labconco Corporation Produits
Click to view available options
Plage de températures (métrique):
-40°C à +40°C
Conditionnement:
1 unité
Description
- A pneumatic mechanism stoppers containers under vacuum on three large adjustable processing shelves, 16.5 x 12 in. each.
- An HCFC/CFC-free refrigeration system and electric heater cool and heat fluid medium that is circulated through the shelves to maintain shelf temperatures within ±1° C.
- Lyo-Works™ OS lets the user set up to 30 programs, with up to 16 steps each; record, chart and store data; and receive email alerts.
- The tray dryer connects to a FreeZone Console Freeze Dryer that provides a real time display of the tray dryer’s parameters on a large capacitive touchscreen.
- For use with any FreeZone Freeze Dryer manufactured after July 2017.
Spécification
Spécification
| Type | Stoppering Tray Dryer |
| Nbre d’étagères | 3 |
| Plage de températures (métrique) | -40°C à +40°C |
| Matériau | Acier inoxydable |
| Comprend | Isolation Valve, 6-Port Manifold |
| Type de bouchon | All except North America |
| Tension | 230 V |