missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FLT3 Monoclonal Antibody (BV10A4), PE, Invitrogen™
Mouse Monoclonal Antibody
Marque: Invitrogen MA110090
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
This antibody will not cross-react with mouse.
FLT3 is a tyrosine protein kinase that is a member of the growth factor receptor tyrosine kinase family. The receptor is activated by binding of the fms-related tyrosine kinase 3 ligand to the extracellular domain, which induces homodimer formation in the plasma membrane leading to autophosphorylation of the receptor. The activated receptor kinase subsequently phosphorylates and activates multiple cytoplasmic effector molecules in pathways involved in apoptosis, proliferation, and differentiation of hematopoietic cells in bone marrow. Mutations that result in the constitutive activation of this receptor result in acute myeloid leukemia and acute lymphoblastic leukemia.Spécification
FLT3 | |
Monoclonal | |
PE | |
FLT3 | |
B230315G04; CD135; CD135 antigen; fetal liver kinase 2; Fetal liver kinase-2; FL cytokine receptor; FLK2; Flk-2; FLT3; Flt-3; fms related tyrosine kinase 3; FMS-like tyrosine kinase 3; fms-related tyrosine kinase 3; growth factor receptor tyrosine kinase type III; Ly72; OTTHUMP00000214829; Receptor-type tyrosine-protein kinase FLT3; RP11-153M24.3; stem cell tyrosine kinase 1; STK1; STK-1; tyrosine-protein kinase FLT3; tyrosine-protein kinase receptor flk-2; wmfl | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
2322 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
BV10A4 | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
P36888 | |
FLT3 | |
BV-173 leukemic cell line | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu