Learn More
Proteintech FlexAble HRP Antibody Labeling Kit for Human IgG
FlexAble HRP Antibody Labeling Kit for Human IgG is a novel antibody labeling kit that uses an affinity linker to conjugate HRP in any buffer condition to Human IgG primary antibodies from any supplier.
Marque: Proteintech KFA110-50
Informations supplémentaires : Poids : 0.02000kg
Beschreibung
FlexAble HRP Antibody Labeling Kit for Human IgG is a novel antibody labeling kit that uses an affinity linker to conjugate HRP in any buffer condition to Human IgG primary antibodies from any supplier. One labeling reaction requires only 0.5 μg of antibody - regardless of the antibody concentration. Label your Human IgG antibody in<10 min without any additional equipment.
Spezifikation
50 Tests | |
Liquid |