Learn More
Proteintech™ FlexAble CoraLite™ Plus 750 Antibody Labeling Kit for Mouse IgG1
This Antibody Labeling Kit conjugates CoraLite™ Plus 750 into mouse IgG1 primary antibodies.
Marque: Proteintech™ KFA024-50
Informations supplémentaires : Poids : 0.00010kg
Description
FlexAble CoraLite™ Plus 750 Antibody Labeling Kit for Mouse IgG1 is a novel antibody labeling kit that uses an affinity linker to conjugate CoraLite™ Plus 750 in any buffer condition to mouse IgG1 primary antibodies from any supplier.
- One labeling reaction requires only 0.5 μg of antibody, regardless of the antibody concentration
- Label your mouse IgG1 antibody in <10 min without any additional equipment
Spécification
FlexLinker CoraLite™ Plus 750 de 50 μl pour IgG1 de souris, FlexQuencher de 100 μl pour IgG1 de souris, FlexBuffer de 500 μl, Manuel, Aimant de protocole | |
1 an à -20°C ou 6 mois à +4°C, éviter la lumière | |
IgG1 |
50 x 0,5 μg Réactions | |
755 nm/780 nm | |
Souris |