Learn More
Proteintech™ FlexAble CoraLite™ 488 Antibody Labeling Kit for Mouse IgG1
This Antibody Labeling Kit conjugates CoraLite™ 488 into mouse IgG1 primary antibodies.
Marque:  Proteintech™ KFA021-200
Informations supplémentaires : Poids : 0.00010kg
Code produit. 17771519
  • 889.00€ / 200 réactions
Expédition estimée: 12-09-2024
Description
FlexAble CoraLite™ 488 Antibody Labeling Kit for Mouse IgG1 is a novel antibody labeling kit that uses an affinity linker to conjugate CoraLite™ 488 in any buffer condition to mouse IgG1 primary antibodies from any supplier.
  • One labeling reaction requires only 0.5 μg of antibody, regardless of the antibody concentration
  • Label your mouse IgG1 antibody in <10 min without any additional equipment
Spécification
Spécification
4 FlexLinker CoraLite™ 488 de 50 μl pour IgG1 de souris, 4 FlexQuencher de 100 μl pour IgG1 de souris, 4 FlexBuffer de 500 μl, Manuel, Aimant de protocole
1 an à -20°C ou 6 mois à +4°C, éviter la lumière
IgG1
200 réactions de 0,5 μl
500 nm/520 nm
Souris