missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Isotemp™ Digital and Analog Dry Bath Incubators

Code produit. 11830740
Click to view available options
Dimensions (L x l x H):
14.2 x 10.8 x 3.3 in. (36.1 x 27.4 x 9.1 cm)
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Includes

Digital Models come with temperature probe, line cord and plug. Analog Models include high and low thermostat, power switch, pilot light, line cord and plug.

Requires

Heating blocks, sold separately, hold regular tubes, microcentrifuge tubes, square cuvets, or microplates.

Provide controlled dry heat for a wide variety of clinical and general chemistry applications

Choice of 1- to 6-block Analog Models, 2- or 4-block Digital Models.

  • Exceptional temperature control and uniformity
  • Temperature range of 5°C above ambient to 130°C
  • Powder-coated steel body construction provides durability for demanding applications
  • Compact design saves valuable bench space
  • Choice of interchangeable heating blocks

Digital Dry Baths with Microprocessor Controller

  • PID microprocessor control, digital temperature setting
  • Just press up/down arrows to set temperature in 0.1° increments
  • Effectively hold samples at constant, warm temperature
  • 5°C above ambient to 130°C range; ±0.4° uniformity at 37°C; stability, ±0.2°C
  • Bright LED display continuously shows actual temperature in °C
  • Temperature probe in bottom of bath provides improved accuracy and control; free probe included
  • Model 125DQ holds two heating blocks; Model 145DQ holds four heating blocks in 2 × 2 configuration
Analog Dry Baths with Dual-thermostat Controls
  • Sloped control panel makes controls highly visible
  • Dual bimetallic thermostat controls
  • Stability is ±1.5°C, uniformity ±0.5°C at 37°C
  • Use thermometer to read current temperature of blocks, then adjust control to reach desired setpoint
  • Efficient design: operate at substantially lower external temperature than other models

Applications:

Ideal for enzyme reactions, inactivation of sera, Rh studies, blood crossmatching and cholesterol determinations

Informations sur la commande

All models accept 2.91L x 3.75W x 2 in.H (7.4 x 9.5 x 5.1cm) blocks, sold separately.

Certifications

Digital Models are CSA approved. Analog Models are UL listed.

Garantie

One year

†International model

TRUSTED_SUSTAINABILITY

Spécification

Matériau Acier
Plage de températures (métrique) Ambient +5° to 130°C
Comprend Microprocessor controller, temperature probe, line cord and plug, holds two heating blocks
Dimensions (L x l x H) 14.2 x 10.8 x 3.3 in. (36.1 x 27.4 x 9.1 cm)
Longueur (impériale) 14.2 in.
Longueur (métrique) 36.1 cm
Largeur (impériale) 10.8 in.
Largeur (métrique) 27.4 cm
Hauteur (impériale) 3.3 in.
Hauteur (métrique) 9.1 cm
Alimentation électrique 240 V, 50/60 Hz
Tension 240 V
Fréquence 50/60 Hz
Afficher plus Afficher moins
Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis